Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R659 aHu CD7 Anti

Catalog No. 5000087260
Encompass_Preferred

Product Name: cFluor R659 Anti-Human CD7 (CD7-6B7)
Product Class: Antibody
Type: Primary
Product Name: cFluor R659 Anti-Human CD7 (CD7-6B7)
Cytek Catalog Number: R7-20327
Marker: CD7
Target Species: Human
Clonality: Monoclonal
Clone: CD7-6B7
Host Species: Mouse
Isotype: IgG2a
Conjugation: cFluor R659
Size: 100 Tests

Catalog No. 50-000-87260 Supplier Cytek Biosciences Inc Supplier No. R720327
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R659 Anti-Human CD7 (CD7-6B7)
Product Class: Antibody
Type: Primary
Product Name: cFluor R659 Anti-Human CD7 (CD7-6B7)
Cytek Catalog Number: R7-20327
Marker: CD7
Target Species: Human
Clonality: Monoclonal
Clone: CD7-6B7
Host Species: Mouse
Isotype: IgG2a
Conjugation: cFluor R659
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.