Learn More
Cytek Biosciences Inc cFluor R659 aHu LOX-1 Anti
Supplier: Cytek Biosciences Inc R720462
Product Name: cFluor R659 Anti-Human LOX-1 (15C4)
Product Class: Antibody
Type: Primary
Product Name: cFluor R659 Anti-Human LOX-1 (15C4)
Cytek Catalog Number: R7-20462
Marker: LOX-1
Target Species: Human
Clonality: Monoclonal
Clone: 15C4
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor R659
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.