Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R668 aHu CD1c Anti

Supplier:  Cytek Biosciences Inc R720120

Encompass_Preferred

Product Name: cFluor R668 Anti-Human CD1c (L161)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD1c (L161)
Cytek Catalog Number: R7-20120
Marker: CD1c
Target Species: Human
Clonality: Monoclonal
Clone: L161
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 25 Tests

Catalog No. 50-000-86999


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.