Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R668 aHu CD314 Anti

Catalog No. 5000087113
Encompass_Preferred

Product Name: cFluor R668 Anti-Human CD314 (NKG2D) (1D11)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD314 (NKG2D) (1D11)
Cytek Catalog Number: R7-20471
Marker: CD314 (NKG2D)
Target Species: Human
Reported Cross Reactivity: African Green|Baboon|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: 1D11
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 100 Tests

Catalog No. 50-000-87113 Supplier Cytek Biosciences Inc Supplier No. R720471
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R668 Anti-Human CD314 (NKG2D) (1D11)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD314 (NKG2D) (1D11)
Cytek Catalog Number: R7-20471
Marker: CD314 (NKG2D)
Target Species: Human
Reported Cross Reactivity: African Green|Baboon|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: 1D11
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.