Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R668 aHu CD8 Anti

Catalog No. 5000086735
Encompass_Preferred

Product Name: cFluor R668 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20228
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 25 Tests

Catalog No. 50-000-86735 Supplier Cytek Biosciences Inc Supplier No. R720228
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R668 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20228
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.