Learn More
Cytek Biosciences Inc cFluor R668 aHu CD8 Anti
Supplier: Cytek Biosciences Inc R720227
Product Name: cFluor R668 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor R668 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20227
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R668
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.