Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R685 aHu CD11c Anti

Catalog No. 5000086917
Encompass_Preferred

Product Name: cFluor R685 Anti-Human CD11c (S-HCL-3)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human CD11c (S-HCL-3)
Cytek Catalog Number: R7-20588
Marker: CD11c
Target Species: Human
Clonality: Monoclonal
Clone: S-HCL-3
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor R685
Size: 25 Tests

Catalog No. 50-000-86917 Supplier Cytek Biosciences Inc Supplier No. R720588
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R685 Anti-Human CD11c (S-HCL-3)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human CD11c (S-HCL-3)
Cytek Catalog Number: R7-20588
Marker: CD11c
Target Species: Human
Clonality: Monoclonal
Clone: S-HCL-3
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor R685
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.