Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R685 aHu CD2 Anti

Catalog No. 5000087176
Encompass_Preferred

Product Name: cFluor R685 Anti-Human CD2 (RPA-2.10)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human CD2 (RPA-2.10)
Cytek Catalog Number: R7-20366
Marker: CD2
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Capuchin Monkey|Chimpanzee|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Pig
Clonality: Monoclonal
Clone: RPA-2.10
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R685
Size: 25 Tests

Catalog No. 50-000-87176 Supplier Cytek Biosciences Inc Supplier No. R720366
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R685 Anti-Human CD2 (RPA-2.10)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human CD2 (RPA-2.10)
Cytek Catalog Number: R7-20366
Marker: CD2
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Capuchin Monkey|Chimpanzee|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Pig
Clonality: Monoclonal
Clone: RPA-2.10
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R685
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.