Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R685 aHu Ig Kappa Anti

Catalog No. 5000086801
Encompass_Preferred

Product Name: cFluor R685 Anti-Human Ig Kappa (Polyclonal)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human Ig Kappa (Polyclonal)
Cytek Catalog Number: R7-20736
Marker: Ig Kappa
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Isotype: IgG
Conjugation: cFluor R685
Size: 25 Tests

Catalog No. 50-000-86801 Supplier Cytek Biosciences Inc Supplier No. R720736
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R685 Anti-Human Ig Kappa (Polyclonal)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human Ig Kappa (Polyclonal)
Cytek Catalog Number: R7-20736
Marker: Ig Kappa
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Isotype: IgG
Conjugation: cFluor R685
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.