Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R685 aMu CD11b Anti

Catalog No. 5000086989
Encompass_Preferred

Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Cytek Catalog Number: R7-20548
Marker: CD11b
Target Species: Human|Mouse
Clonality: Monoclonal
Clone: M1/70
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor R685
Size: 25 Tests

Catalog No. 50-000-86989 Supplier Cytek Biosciences Inc Supplier No. R720548
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Cytek Catalog Number: R7-20548
Marker: CD11b
Target Species: Human|Mouse
Clonality: Monoclonal
Clone: M1/70
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor R685
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.