Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R685 aMu CD11b Anti

Supplier:  Cytek Biosciences Inc R720547

Encompass_Preferred

Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Product Class: Antibody
Type: Primary
Product Name: cFluor R685 Anti-Human/Mouse CD11b (M1/70)
Cytek Catalog Number: R7-20547
Marker: CD11b
Target Species: Human|Mouse
Clonality: Monoclonal
Clone: M1/70
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor R685
Size: 100 Tests

Catalog No. 50-000-87298


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.