Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R720 aHu CD161 Anti

Catalog No. 5000087270
Encompass_Preferred

Product Name: cFluor R720 Anti-Human CD161 (NK1.1) (HP-3G10)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD161 (NK1.1) (HP-3G10)
Cytek Catalog Number: R7-20385
Marker: CD161 (NK1.1)
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Rhesus Macaque
Clonality: Monoclonal
Clone: HP-3G10
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R720
Size: 100 Tests

Catalog No. 50-000-87270 Supplier Cytek Biosciences Inc Supplier No. R720385
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R720 Anti-Human CD161 (NK1.1) (HP-3G10)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD161 (NK1.1) (HP-3G10)
Cytek Catalog Number: R7-20385
Marker: CD161 (NK1.1)
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Rhesus Macaque
Clonality: Monoclonal
Clone: HP-3G10
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R720
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.