Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R720 aHu CD5 Anti

Catalog No. 5000086825
Encompass_Preferred

Product Name: cFluor R720 Anti-Human CD5 (L17F12)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD5 (L17F12)
Cytek Catalog Number: R7-20096
Marker: CD5
Target Species: Human
Clonality: Monoclonal
Clone: L17F12
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor R720
Size: 25 Tests

Catalog No. 50-000-86825 Supplier Cytek Biosciences Inc Supplier No. R720096
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R720 Anti-Human CD5 (L17F12)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD5 (L17F12)
Cytek Catalog Number: R7-20096
Marker: CD5
Target Species: Human
Clonality: Monoclonal
Clone: L17F12
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor R720
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.