Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R720 aHu CD58 Anti

Catalog No. 5000087014
Encompass_Preferred

Product Name: cFluor R720 Anti-Human CD58 (MEM-63)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD58 (MEM-63)
Cytek Catalog Number: R7-20701
Marker: CD58
Target Species: Human
Clonality: Monoclonal
Clone: MEM-63
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor R720
Size: 100 Tests

Catalog No. 50-000-87014 Supplier Cytek Biosciences Inc Supplier No. R720701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R720 Anti-Human CD58 (MEM-63)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD58 (MEM-63)
Cytek Catalog Number: R7-20701
Marker: CD58
Target Species: Human
Clonality: Monoclonal
Clone: MEM-63
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor R720
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.