Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R720 aHu CD66b Anti

Supplier:  Cytek Biosciences Inc R720331

Encompass_Preferred

Product Name: cFluor R720 Anti-Human CD66b (G10F5)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human CD66b (G10F5)
Cytek Catalog Number: R7-20331
Marker: CD66b
Target Species: Human
Clonality: Monoclonal
Clone: G10F5
Host Species: Mouse
Isotype: IgM, k
Conjugation: cFluor R720
Size: 100 Tests

Catalog No. 50-000-87126


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.