Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R780 aHu CD123 Anti

Catalog No. 5000087400
Encompass_Preferred

Product Name: cFluor R780 Anti-Human CD123 (6H6)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Human CD123 (6H6)
Cytek Catalog Number: R7-20738
Marker: CD123
Target Species: Human
Reported Cross Reactivity: Rhesus Macaque
Clonality: Monoclonal
Clone: 6H6
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R780
Size: 25 Tests

Catalog No. 50-000-87400 Supplier Cytek Biosciences Inc Supplier No. R720738
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor R780 Anti-Human CD123 (6H6)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Human CD123 (6H6)
Cytek Catalog Number: R7-20738
Marker: CD123
Target Species: Human
Reported Cross Reactivity: Rhesus Macaque
Clonality: Monoclonal
Clone: 6H6
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R780
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.