Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R780 aHu CD56 Anti

Catalog No. 5000087275
Encompass_Preferred

Product Name: cFluor R780 Anti-Human CD56 (5.1H11)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Human CD56 (5.1H11)
Cytek Catalog Number: R7-20710
Marker: CD56
Target Species: Human
Clonality: Monoclonal
Clone: 5.1H11
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R780
Size: 25 Tests

Catalog No. 50-000-87275 Supplier Cytek Biosciences Inc Supplier No. R720710
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R780 Anti-Human CD56 (5.1H11)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Human CD56 (5.1H11)
Cytek Catalog Number: R7-20710
Marker: CD56
Target Species: Human
Clonality: Monoclonal
Clone: 5.1H11
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R780
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.