Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R780 aMu TCR Anti

Catalog No. 5000087155
Encompass_Preferred

Product Name: cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)
Cytek Catalog Number: R7-20558
Marker: TCR γ/δ
Target Species: Mouse
Clonality: Monoclonal
Clone: eBio GL3
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor R780
Size: 25 Tests

Catalog No. 50-000-87155 Supplier Cytek Biosciences Inc Supplier No. R720558
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)
Product Class: Antibody
Type: Primary
Product Name: cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)
Cytek Catalog Number: R7-20558
Marker: TCR γ/δ
Target Species: Mouse
Clonality: Monoclonal
Clone: eBio GL3
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor R780
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.