Learn More
Cytek Biosciences Inc CFLUOR R780 ANTI MOUS 100TESTS
Catalog No. NC3257591
Encompass_Preferred
cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)Clone: eBio GL3Reactivity: MouseFormat: cFluor® R7801Isotype: Armenian Hamster IgGTested Dilution: 5 μL / testApplication: Flow CytometryStorage: 2-8°C and protected from Light. Do not freezeFormulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% BSA (Origin USA)
Catalog No. NC3257591 Supplier Cytek Biosciences Inc Supplier No. R720557
cFluor R780 Anti-Mouse TCR γ/δ (eBio GL3)Clone: eBio GL3Reactivity: MouseFormat: cFluor® R7801Isotype: Armenian Hamster IgGTested Dilution: 5 μL / testApplication: Flow CytometryStorage: 2-8°C and protected from Light. Do not freezeFormulation: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% BSA (Origin USA)