Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R840 aHu CD8 Anti

Supplier:  Cytek Biosciences Inc R720235

Encompass_Preferred

Product Name: cFluor R840 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor R840 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20235
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor R840
Size: 100 Tests

Catalog No. 50-000-86820


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.