Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor UV515 aMu CD25 Anti

Catalog No. 5000086558
Encompass_Preferred

Product Name: cFluor UV515 Anti-Mouse CD25 (PC61.5.3)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV515 Anti-Mouse CD25 (PC61.5.3)
Cytek Catalog Number: R7-20865
Marker: CD25
Target Species: Mouse
Clonality: Monoclonal
Clone: PC61.5.3
Host Species: Rat
Isotype: IgG1, l
Conjugation: cFluor UV515
Size: 100 Tests

Catalog No. 50-000-86558 Supplier Cytek Biosciences Inc Supplier No. R720865
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor UV515 Anti-Mouse CD25 (PC61.5.3)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV515 Anti-Mouse CD25 (PC61.5.3)
Cytek Catalog Number: R7-20865
Marker: CD25
Target Species: Mouse
Clonality: Monoclonal
Clone: PC61.5.3
Host Species: Rat
Isotype: IgG1, l
Conjugation: cFluor UV515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.