Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor UV610 aMu CD4 Anti

Catalog No. 5000086519
Encompass_Preferred

Product Name: cFluor UV610 Anti-Mouse CD4 (RM4-5)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV610 Anti-Mouse CD4 (RM4-5)
Cytek Catalog Number: R7-20923
Marker: CD4
Target Species: Mouse
Clonality: Monoclonal
Clone: RM4-5
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor UV610
Size: 100 Tests

Catalog No. 50-000-86519 Supplier Cytek Biosciences Inc Supplier No. R720923
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor UV610 Anti-Mouse CD4 (RM4-5)
Product Class: Antibody
Type: Primary
Product Name: cFluor UV610 Anti-Mouse CD4 (RM4-5)
Cytek Catalog Number: R7-20923
Marker: CD4
Target Species: Mouse
Clonality: Monoclonal
Clone: RM4-5
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor UV610
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.