Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V420 aHu CD8 Anti

Catalog No. 5000087206
Encompass_Preferred

Product Name: cFluor V420 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor V420 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20238
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V420
Size: 25 Tests

Catalog No. 50-000-87206 Supplier Cytek Biosciences Inc Supplier No. R720238
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V420 Anti-Human CD8 (SK1)
Product Class: Antibody
Type: Primary
Product Name: cFluor V420 Anti-Human CD8 (SK1)
Cytek Catalog Number: R7-20238
Marker: CD8
Target Species: Human
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque
Clonality: Monoclonal
Clone: SK1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V420
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.