Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V435 aHu CD200 Anti

Supplier:  Cytek Biosciences Inc R720755

Encompass_Preferred

Product Name: cFluor V435 Anti-Human CD200 (OX-104)
Product Class: Antibody
Type: Primary
Product Name: cFluor V435 Anti-Human CD200 (OX-104)
Cytek Catalog Number: R7-20755
Marker: CD200
Target Species: Human
Clonality: Monoclonal
Clone: OX-104
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V435
Size: 100 Tests

Catalog No. 50-000-87055


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.