Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V435 aHu CD56 Anti

Catalog No. 5000087384
Encompass_Preferred

Product Name: cFluor V435 Anti-Human CD56 (B-A19)
Product Class: Antibody
Type: Primary
Product Name: cFluor V435 Anti-Human CD56 (B-A19)
Cytek Catalog Number: R7-21395
Marker: CD56
Target Species: Human
Clonality: Monoclonal
Clone: B-A19
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V435
Size: 100 Tests

Catalog No. 50-000-87384 Supplier Cytek Biosciences Inc Supplier No. R721395
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V435 Anti-Human CD56 (B-A19)
Product Class: Antibody
Type: Primary
Product Name: cFluor V435 Anti-Human CD56 (B-A19)
Cytek Catalog Number: R7-21395
Marker: CD56
Target Species: Human
Clonality: Monoclonal
Clone: B-A19
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V435
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.