Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD123 Anti

Supplier:  Cytek Biosciences Inc R720192

Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD123 (6H6)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD123 (6H6)
Cytek Catalog Number: R7-20192
Marker: CD123
Target Species: Human
Reported Cross Reactivity: Rhesus Macaque
Clonality: Monoclonal
Clone: 6H6
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 25 Tests

Catalog No. 50-000-86775


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.