Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD16 Anti

Catalog No. 5000087207
Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD16 (3G8)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD16 (3G8)
Cytek Catalog Number: R7-20183
Marker: CD16
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Common Marmoset|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Sooty Mangabey|Squirrel Monkey
Clonality: Monoclonal
Clone: 3G8
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 100 Tests

Catalog No. 50-000-87207 Supplier Cytek Biosciences Inc Supplier No. R720183
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V450 Anti-Human CD16 (3G8)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD16 (3G8)
Cytek Catalog Number: R7-20183
Marker: CD16
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Common Marmoset|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Sooty Mangabey|Squirrel Monkey
Clonality: Monoclonal
Clone: 3G8
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.