Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD19 Anti

Catalog No. 5000087154
Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD19 (HIB19)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD19 (HIB19)
Cytek Catalog Number: R7-20484
Marker: CD19
Target Species: Human
Reported Cross Reactivity: Chimpanzee|Rhesus Macaque
Clonality: Monoclonal
Clone: HIB19
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 25 Tests

Catalog No. 50-000-87154 Supplier Cytek Biosciences Inc Supplier No. R720484
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V450 Anti-Human CD19 (HIB19)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD19 (HIB19)
Cytek Catalog Number: R7-20484
Marker: CD19
Target Species: Human
Reported Cross Reactivity: Chimpanzee|Rhesus Macaque
Clonality: Monoclonal
Clone: HIB19
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.