Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD48 Anti

Catalog No. 5000086527
Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD48 (MEM-102)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD48 (MEM-102)
Cytek Catalog Number: R7-20772
Marker: CD48
Target Species: Human
Clonality: Monoclonal
Clone: MEM-102
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor V450
Size: 25 Tests

Catalog No. 50-000-86527 Supplier Cytek Biosciences Inc Supplier No. R720772
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V450 Anti-Human CD48 (MEM-102)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD48 (MEM-102)
Cytek Catalog Number: R7-20772
Marker: CD48
Target Species: Human
Clonality: Monoclonal
Clone: MEM-102
Host Species: Mouse
Isotype: IgG1
Conjugation: cFluor V450
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.