Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD9 Anti

Supplier:  Cytek Biosciences Inc R720682

Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Cytek Catalog Number: R7-20682
Marker: CD9
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Bovine|Cynomolgus Monkey|Dog|Equine|Rabbit|Rhesus Macaque|Ovine
Clonality: Monoclonal
Clone: HI9a
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 25 Tests

Catalog No. 50-000-87074


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.