Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V450 aHu CD9 Anti

Catalog No. 5000087315
Encompass_Preferred

Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Cytek Catalog Number: R7-20681
Marker: CD9
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Bovine|Cynomolgus Monkey|Dog|Equine|Rabbit|Rhesus Macaque|Ovine
Clonality: Monoclonal
Clone: HI9a
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 100 Tests

Catalog No. 50-000-87315 Supplier Cytek Biosciences Inc Supplier No. R720681
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Product Class: Antibody
Type: Primary
Product Name: cFluor V450 Anti-Human CD9 (HI9a)
Cytek Catalog Number: R7-20681
Marker: CD9
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Bovine|Cynomolgus Monkey|Dog|Equine|Rabbit|Rhesus Macaque|Ovine
Clonality: Monoclonal
Clone: HI9a
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V450
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.