Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V505 aMu CD161 Anti

Catalog No. 5000086992
Encompass_Preferred

Product Name: cFluor V505 Anti-Mouse CD161 (NK1.1) (PK136)
Product Class: Antibody
Type: Primary
Product Name: cFluor V505 Anti-Mouse CD161 (NK1.1) (PK136)
Cytek Catalog Number: R7-20568
Marker: CD161 (NK1.1)
Target Species: Mouse
Clonality: Monoclonal
Clone: PK136
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V505
Size: 25 Tests

Catalog No. 50-000-86992 Supplier Cytek Biosciences Inc Supplier No. R720568
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V505 Anti-Mouse CD161 (NK1.1) (PK136)
Product Class: Antibody
Type: Primary
Product Name: cFluor V505 Anti-Mouse CD161 (NK1.1) (PK136)
Cytek Catalog Number: R7-20568
Marker: CD161 (NK1.1)
Target Species: Mouse
Clonality: Monoclonal
Clone: PK136
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V505
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.