Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V515 aHu CD14 Anti

Supplier:  Cytek Biosciences Inc R721067

Encompass_Preferred

Product Name: cFluor V515 Anti-Human CD14 (TÜK4)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Human CD14 (TÜK4)
Cytek Catalog Number: R7-21067
Marker: CD14
Target Species: Human
Reported Cross Reactivity: Non-Human Primate|Rhesus Macaque|Cynomolgus Monkey|Pigtail Monkey|Cotton-Topped Tamarin|Pig|Bovine|Ovine|Goat|Dog|Mink|Rabbit
Clonality: Monoclonal
Clone: TÜK4
Host Species: Mouse
Isotype: IgG2a
Conjugation: cFluor V515
Size: 100 Tests

Catalog No. 50-000-86875


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.