Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V515 aHu CD69 Anti

Catalog No. 5000086960
Encompass_Preferred

Product Name: cFluor V515 Anti-Human CD69 (FN50)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Human CD69 (FN50)
Cytek Catalog Number: R7-21081
Marker: CD69
Target Species: Human
Clonality: Monoclonal
Clone: FN50
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V515
Size: 100 Tests

Catalog No. 50-000-86960 Supplier Cytek Biosciences Inc Supplier No. R721081
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V515 Anti-Human CD69 (FN50)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Human CD69 (FN50)
Cytek Catalog Number: R7-21081
Marker: CD69
Target Species: Human
Clonality: Monoclonal
Clone: FN50
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.