Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V515 aHu MHCI Anti

Catalog No. 5000086970
Encompass_Preferred

Product Name: cFluor V515 Anti-Human MHC Class I (HLA-ABC) (W6/32)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Human MHC Class I (HLA-ABC) (W6/32)
Cytek Catalog Number: R7-21083
Marker: MHC Class I (HLA-ABC)
Target Species: Human
Clonality: Monoclonal
Clone: W6/32
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V515
Size: 100 Tests

Catalog No. 50-000-86970 Supplier Cytek Biosciences Inc Supplier No. R721083
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V515 Anti-Human MHC Class I (HLA-ABC) (W6/32)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Human MHC Class I (HLA-ABC) (W6/32)
Cytek Catalog Number: R7-21083
Marker: MHC Class I (HLA-ABC)
Target Species: Human
Clonality: Monoclonal
Clone: W6/32
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.