Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V515 aMu CD19 Anti

Catalog No. 5000087061
Encompass_Preferred

Product Name: cFluor V515 Anti-Mouse CD19 (6D5)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Mouse CD19 (6D5)
Cytek Catalog Number: R7-21087
Marker: CD19
Target Species: Mouse
Clonality: Monoclonal
Clone: 6D5
Host Species: Rat
Isotype: IgG2a
Conjugation: cFluor V515
Size: 100 Tests

Catalog No. 50-000-87061 Supplier Cytek Biosciences Inc Supplier No. R721087
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V515 Anti-Mouse CD19 (6D5)
Product Class: Antibody
Type: Primary
Product Name: cFluor V515 Anti-Mouse CD19 (6D5)
Cytek Catalog Number: R7-21087
Marker: CD19
Target Species: Mouse
Clonality: Monoclonal
Clone: 6D5
Host Species: Rat
Isotype: IgG2a
Conjugation: cFluor V515
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.