Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V547 aHu CD20 Anti

Catalog No. 5000087044
Encompass_Preferred

Product Name: cFluor V547 Anti-Human CD20 (2H7)
Product Class: Antibody
Type: Primary
Product Name: cFluor V547 Anti-Human CD20 (2H7)
Cytek Catalog Number: R7-20112
Marker: CD20
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Squirrel Monkey
Clonality: Monoclonal
Clone: 2H7
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor V547
Size: 25 Tests

Catalog No. 50-000-87044 Supplier Cytek Biosciences Inc Supplier No. R720112
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V547 Anti-Human CD20 (2H7)
Product Class: Antibody
Type: Primary
Product Name: cFluor V547 Anti-Human CD20 (2H7)
Cytek Catalog Number: R7-20112
Marker: CD20
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Squirrel Monkey
Clonality: Monoclonal
Clone: 2H7
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor V547
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.