Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V547 aHu HLA-DR Anti

Supplier:  Cytek Biosciences Inc R720724

Encompass_Preferred

Product Name: cFluor V547 Anti-Human HLA-DR (L243)
Product Class: Antibody
Type: Primary
Product Name: cFluor V547 Anti-Human HLA-DR (L243)
Cytek Catalog Number: R7-20724
Marker: HLA-DR
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Chimpanzee|Cynomolgus Monkey|Dog|Common Marmoset|Rhesus Macaque|Squirrel Monkey
Clonality: Monoclonal
Clone: L243
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V547
Size: 25 Tests

Catalog No. 50-000-87199


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.