Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V547 aMu CD45 Anti

Supplier:  Cytek Biosciences Inc R720572

Encompass_Preferred

Product Name: cFluor V547 Anti-Mouse CD45 (30-F11)
Product Class: Antibody
Type: Primary
Product Name: cFluor V547 Anti-Mouse CD45 (30-F11)
Cytek Catalog Number: R7-20572
Marker: CD45
Target Species: Mouse
Clonality: Monoclonal
Clone: 30-F11
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor V547
Size: 25 Tests

Catalog No. 50-000-86994


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.