Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V570 aHu CD33 Anti

Catalog No. 5000086569
Encompass_Preferred

Product Name: cFluor V570 Anti-Human CD33 (WM53)
Product Class: Antibody
Type: Primary
Product Name: cFluor V570 Anti-Human CD33 (WM53)
Cytek Catalog Number: R7-21103
Marker: CD33
Target Species: Human
Clonality: Monoclonal
Clone: WM53
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V570
Size: 100 Tests

Catalog No. 50-000-86569 Supplier Cytek Biosciences Inc Supplier No. R721103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V570 Anti-Human CD33 (WM53)
Product Class: Antibody
Type: Primary
Product Name: cFluor V570 Anti-Human CD33 (WM53)
Cytek Catalog Number: R7-21103
Marker: CD33
Target Species: Human
Clonality: Monoclonal
Clone: WM53
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V570
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.