Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V570 aMu CD45R Anti

Catalog No. 5000086530
Encompass_Preferred

Product Name: cFluor V570 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Product Class: Antibody
Type: Primary
Product Name: cFluor V570 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Cytek Catalog Number: R7-21121
Marker: CD45R (B220)
Target Species: Human|Mouse
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RA3-6B2
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor V570
Size: 100 Tests

Catalog No. 50-000-86530 Supplier Cytek Biosciences Inc Supplier No. R721121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V570 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Product Class: Antibody
Type: Primary
Product Name: cFluor V570 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Cytek Catalog Number: R7-21121
Marker: CD45R (B220)
Target Species: Human|Mouse
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RA3-6B2
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor V570
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.