Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V605 aMu CD161 Anti

Catalog No. 5000086956
Encompass_Preferred

Product Name: cFluor V605 Anti-Mouse CD161 (NK1.1) (PK136)
Product Class: Antibody
Type: Primary
Product Name: cFluor V605 Anti-Mouse CD161 (NK1.1) (PK136)
Cytek Catalog Number: R7-21141
Marker: CD161 (NK1.1)
Target Species: Mouse
Clonality: Monoclonal
Clone: PK136
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V605
Size: 100 Tests

Catalog No. 50-000-86956 Supplier Cytek Biosciences Inc Supplier No. R721141
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V605 Anti-Mouse CD161 (NK1.1) (PK136)
Product Class: Antibody
Type: Primary
Product Name: cFluor V605 Anti-Mouse CD161 (NK1.1) (PK136)
Cytek Catalog Number: R7-21141
Marker: CD161 (NK1.1)
Target Species: Mouse
Clonality: Monoclonal
Clone: PK136
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor V605
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.