Learn More
Cytek Biosciences Inc cFluor V610 aHu CD20 Anti
Supplier: Cytek Biosciences Inc R720669
Product Name: cFluor V610 Anti-Human CD20 (2H7)
Product Class: Antibody
Type: Primary
Product Name: cFluor V610 Anti-Human CD20 (2H7)
Cytek Catalog Number: R7-20669
Marker: CD20
Target Species: Human
Reported Cross Reactivity: Baboon|Capuchin Monkey|Chimpanzee|Cynomolgus Monkey|Pigtail Monkey|Rhesus Macaque|Squirrel Monkey
Clonality: Monoclonal
Clone: 2H7
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: cFluor V610
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.