Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V670 aHu CD4 Anti

Catalog No. 5000086586
Encompass_Preferred

Product Name: cFluor V670 Anti-Human CD4 (RPA-T4)
Product Class: Antibody
Type: Primary
Product Name: cFluor V670 Anti-Human CD4 (RPA-T4)
Cytek Catalog Number: R7-21161
Marker: CD4
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RPA-T4
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V670
Size: 100 Tests

Catalog No. 50-000-86586 Supplier Cytek Biosciences Inc Supplier No. R721161
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Product Name: cFluor V670 Anti-Human CD4 (RPA-T4)
Product Class: Antibody
Type: Primary
Product Name: cFluor V670 Anti-Human CD4 (RPA-T4)
Cytek Catalog Number: R7-21161
Marker: CD4
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RPA-T4
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V670
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.