Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V670 aHu Ig Lambda Anti

Catalog No. 5000087006
Encompass_Preferred

Product Name: cFluor V670 Anti-Human Ig Lambda (Polyclonal)
Product Class: Antibody
Type: Primary
Product Name: cFluor V670 Anti-Human Ig Lambda (Polyclonal)
Cytek Catalog Number: R7-20759
Marker: Ig Lambda
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Isotype: IgG
Conjugation: cFluor V670
Size: 100 Tests

Catalog No. 50-000-87006 Supplier Cytek Biosciences Inc Supplier No. R720759
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: cFluor V670 Anti-Human Ig Lambda (Polyclonal)
Product Class: Antibody
Type: Primary
Product Name: cFluor V670 Anti-Human Ig Lambda (Polyclonal)
Cytek Catalog Number: R7-20759
Marker: Ig Lambda
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Isotype: IgG
Conjugation: cFluor V670
Size: 100 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.