Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor V780 aHu CD3 Anti

Supplier:  Cytek Biosciences Inc R720774

Encompass_Preferred

Product Name: cFluor V780 Anti-Human CD3 (SK7)
Product Class: Antibody
Type: Primary
Product Name: cFluor V780 Anti-Human CD3 (SK7)
Cytek Catalog Number: R7-20774
Marker: CD3
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: SK7
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V780
Size: 25 Tests

Catalog No. 50-000-86582


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.