Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor YG600 aHu CD71 Anti

Supplier:  Cytek Biosciences Inc R720729

Encompass_Preferred

Product Name: cFluor YG600 Anti-Human CD71 (OKT9)
Product Class: Antibody
Type: Primary
Product Name: cFluor YG600 Anti-Human CD71 (OKT9)
Cytek Catalog Number: R7-20729
Marker: CD71
Target Species: Human
Clonality: Monoclonal
Clone: OKT9
Host Species: Mouse
Isotype: BALB/c IgG1, k
Conjugation: cFluor YG600
Size: 100 Tests

Catalog No. 50-000-87018


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.