Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cfm 1571 hydrochloride | 1215548-30-3 | 99.5% | 444.95 | C23H29ClN4O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10754610MG

Encompass_Preferred

CFM 1571 hydrochloride is the hydrochloride salt of a small-molecule stimulator of soluble guanylyl cyclase (sGC) intended for laboratory research use. It is provided as a solid material with high purity and characterized identity for biochemical and pharmacological studies.

  • High purity of 99.47%.
  • Molecular weight 444.95 g/mol.
  • Chemical formula C23H29ClN4O3.
  • White to off-white solid appearance.
  • Soluble in DMSO for solution studies.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in milligram-scale quantities for research applications.

Catalog No. 50-002-39860


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.