Learn More
Medchemexpress LLC Cfm 1571 hydrochloride | 1215548-30-3 | 99.5% | 444.95 g·mol⁻¹ | C23H29ClN4O3 | 5 MG
SDP
Catalog No. 5000239881
Encompass_Preferred
CFM 1571 hydrochloride is a hydrochloride salt of a small-molecule soluble guanylate cyclase (sGC) stimulator used for laboratory research. It functions as a biochemical probe to study nitric oxide-cGMP signaling and sGC-mediated pathways. Supplied as a high-purity solid for in vitro and biochemical assays; not for human or animal use.
  • High purity suitable for research applications.
  • Hydrochloride salt for improved solubility in polar solvents.
  • Used to probe sGC and nitric oxide-cGMP signaling pathways.
  • Available in small pack sizes for screening and optimization studies.
  • Characterized by CAS 1215548-30-3 and defined molecular formula.
Catalog No. 50-002-39881 Supplier Medchemexpress LLC Supplier No. HY1075465MG
CFM 1571 hydrochloride is a hydrochloride salt of a small-molecule soluble guanylate cyclase (sGC) stimulator used for laboratory research. It functions as a biochemical probe to study nitric oxide-cGMP signaling and sGC-mediated pathways. Supplied as a high-purity solid for in vitro and biochemical assays; not for human or animal use.
  • High purity suitable for research applications.
  • Hydrochloride salt for improved solubility in polar solvents.
  • Used to probe sGC and nitric oxide-cGMP signaling pathways.
  • Available in small pack sizes for screening and optimization studies.
  • Characterized by CAS 1215548-30-3 and defined molecular formula.