Learn More
Climet Instruments Climet CI-154 Particle Counter, 1 CFM Flow Rate, VHP Compatible, Legacy Model
SDP
Catalog No. NC2328803
Encompass_Preferred
The CI-154 Portable Particle Counter from CLIMET INSTRUMENTS is designed for precise environmental monitoring in cleanrooms and controlled environments. With a flow rate of 1 CFM, it ensures accurate particle measurement across multiple size channels. Its robust data management features and industry compliance make it ideal for pharmaceutical and semiconductor applications. The inclusion of an internal thermal printer and user-defined programs enhances usability, while its durable design promises reliability for years to come.
Catalog No. NC2328803 Supplier Climet Instruments Supplier No. 95015401
Add to Cart
Add to Cart
Request a Quote
The CI-154 Portable Particle Counter from CLIMET INSTRUMENTS is designed for precise environmental monitoring in cleanrooms and controlled environments. With a flow rate of 1 CFM, it ensures accurate particle measurement across multiple size channels. Its robust data management features and industry compliance make it ideal for pharmaceutical and semiconductor applications. The inclusion of an internal thermal printer and user-defined programs enhances usability, while its durable design promises reliability for years to come.